Clam, Gill Slide
470181-666
: Avantor
- Description:Clam, Gill
- Cat. No.:470181-666
- Stain:Hematoxylin and Eosin
- Supplier no.:WARD470181-666
- Orientation:Cross section
Multifunctional Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows typical ciliated epithelium on gill filaments. Absorbs oxygen and separates small non-edible particles from food particles before being digested.