ContactUsLinkComponent
Chymosin, Recombinant Rennin
Chymosin, Recombinant Rennin
  470177-178
 :  
undefined
Chymosin, Recombinant Rennin
  470177-178
 :  R630112
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Chymosin (Recombinant Rennin), 100 mL
  • Cat. No.:
    470177-178
  • Supplier no.:
    WARD470177-178
A 100% natural chymosin from natural fermentation, chymosin is a coagulant often used for cheesemaking. Activity: Minimum 600 International Milk Clotting Units/ml (IMCU/ml). pH: 5.60 to 5.95. Refrigeration is required. Minimum Shelf Life (months) is 12 months. Used in lab 1C of Biotechnology: Science of the New Millennium curriculum.
Perishable. Delivery date required.
 : This product is designed for educational and teaching laboratories and no certificate of analysis is available.