Order Entry
United States
ContactUsLinkComponent
Chromobacterium violaceum
Chromobacterium violaceum
Catalog # 470179-192
Supplier:  Ward's Science
undefined
Chromobacterium violaceum
Catalog # 470179-192
Supplier:  Ward's Science
Supplier Number:  154931A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Chromobacterium violaceum
  • Cat. No.:
    470179-192
  • Packaging:
    Tube
  • Media:
    Nutrient broth
  • Supplier no.:
    WARD470179-192
  • Incubation conditions:
    30 °C

Specifications

About this item

Found in fresh water. Chromogenesis; blue to purple pigments produced. Motile. Rods. Gram negative reaction. Aerobic. Incubation temperature 30°C. Do not refrigerate because it cannot survive in temperatures below 25°C. Nutrient broth growth medium. Note: Pathogen.

Packaged in a 16 x 125 mm tubes. Call our Customer Service for availability of demo plates and other special orders.

  • Biosafety Level 2
  • Canada: Containment Level 2 - PHAC License required