ContactUsLinkComponent
Chilomastix mesnili, Trophozoites Slide
Chilomastix mesnili, Trophozoites Slide
Catalog # 470181-878
Supplier:  Avantor
undefined
Chilomastix mesnili, Trophozoites Slide
Catalog # 470181-878
Supplier:  Avantor
Specifications
  • Description:
    Chilomastix mesnili, Trophozoites
  • Environmentally Preferable:
  • Cat. No.:
    470181-878
  • Supplier no.:
    WARD470181-878
  • Orientation:
    Whole mount
Specifications
About this item
Non-parasitic Gut Microflora
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Common flagellate found in the intestine of man. Life cycle stage when parasite is absorbing nutrients from the host.