ContactUsLinkComponent
Chick Development Slide Set
Chick Development Slide Set
  470177-842
 :  Avantor
undefined
Chick Development Slide Set
  470177-842
 :  Avantor
  • Type:
    Cellular Development
  • Description:
    Chick Development Slide Set
  • No. of slides:
    18
  • Cat. No.:
    470177-842
  • Supplier no.:
    WARD470177-842
Comprehensive Overview of Developing Structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
 : Set includes 12–16-Hour (wm); 18-Hour (wm); 18-Hour (cs); 24-Hour (wm); 24-Hour (cs); 24-Hour (sect); 33-Hour (wm); 33-Hour (cs); 33-Hour (sect); 56-Hour (wm); 56-Hour (cs); 56-Hour (sect); 72-Hour (wm); 72-Hour (cs); 72-Hour (sect); 96-Hour (wm); 96-Hour (cs); and 96-Hour (sect).