Chemistry with Vernier Lab Book
470017-576
- Description:Chemistry with Vernier
- Cat. No.:470017-576
- Supplier no.:CWV
Step-by-Step Experiments That Are Easily Executable
With 36 experiments designed for use with Vernier probeware, this lab book provides a full curriculum of chemistry labs. Attain a comprehensive coverage of topics such as thermochemistry, acid-base reactions, equilibrium, electrochemistry, electrolytes through the included experiment ideas. Each experiment is provided with instructions, a materials list, data tables, and questions, teacher information for each experiment, and word processing files of the student sections on a CD for easy print-out and editing.