ContactUsLinkComponent
Chemiluminescence Student Activity
Chemiluminescence Student Activity
Catalog # 470105-262
Supplier:  Lab-Aids
undefined
Chemiluminescence Student Activity
Catalog # 470105-262
Supplier:  Lab-Aids
Supplier Number:  701A
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Chemiluminescence Student Activity
  • No. of lab groups:
    16
  • Cat. No.:
    470105-262
  • Includes:
    Teacher's Guide, Student Guides and Worksheets, SDS
  • Supplier no.:
    701A
Specifications
About this item
Produce Light Energy from a Chemical Reaction!
  • Observe ratios of chemicals used and effects of temperature on chemiluminescence
  • Demonstrates oxidation-reduction reactions
  • Designed for 16 groups of 2-student teams
  • Requires one 50 minute class period
Chemiluminescence involves a chemical reaction that produces light energy. In this Lab-Aids® kit, students conduct a series of simple, safe and dramatic activities that produce light energy (chemiluminescence) using luminol and an activator solution to create a blue light. Students are introduced to an oxidation-reduction reaction as well as rates of reaction, ratios of chemicals used in the reaction and effects of temperature on chemiluminescence. They will make several observations regarding results of the reaction, and the effects of ratios to solutions and temperature on chemiluminescence.
Ordering information: Materials include Calibrated tubes with caps, Environment cups, Activator Solution, Luminol (Reagent A) powder.