ContactUsLinkComponent
Ward's® Canine Skeleton
  470028-010
 :  Avantor
undefined
Ward's® Canine Skeleton
  470028-010
 :  Avantor
  • Description:
    WARD’S Canine Skeleton
  • Environmentally Preferable:
  • Cat. No.:
    470028-010
  • Supplier no.:
    WARD470028-010
View Typical Skeletal Anatomy of Dogs
Natural bone. The ligamentary skeleton of a medium-size mongrel, Canis familiaris, has been carefully prepared to detail skeletal features common to canines. It is mounted in a normal walking pose on a wooden base and attached by clear plexiglas supports. A key identifying 122 structures is included. Base size: 27 3/4"L x 9"W. Skeleton size: approximately 15"H x 22 1/2"L.