Orders LinkContactUsLinkComponent
Camelid Blood Slide
Camelid Blood Slide
Camelid blood microscope slide, Smear, Giemsa
Catalog # 470182-766
Supplier:   Avantor
Camelid Blood Slide
Catalog # 470182-766
Supplier:  Avantor
Camelid Blood Slide
Camelid Blood Slide
Camelid blood microscope slide, Smear, Giemsa
Price
$5.95
Log in for availability
Each
Product Details & Documents
There is no nucleus in the oval-shaped erythrocytes, which can swell up to 240% of their normal size without bursting. From llama.
Specifications
  • Description:
    Camelid blood microscope slide
  • Stain:
    Giemsa
  • Orientation:
    Smear