Brine Shrimp Eggs
Brine Shrimp Eggs
Catalog # 470152-880
Supplier: Avantor
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Place these eggs in a brine solution for two days in order to make the shrimp (Artemia) hatch. Our special processing technique allows you to store the eggs for nearly indefinite periods of time. 7 grams.
Specifications
- Description:Brine Shrimp Eggs