ContactUsLinkComponent
Ward's® Compact Bone Slide
  470182-656
 :  Avantor
undefined
Ward's® Compact Bone Slide
  470182-656
 :  Avantor
  • Description:
    Bone, Compact Slide
  • Environmentally Preferable:
  • Cat. No.:
    470182-656
  • Stain:
    Hematoxylin and Eosin
  • Supplier no.:
    WARD470182-656
  • Orientation:
    Cross section & longitudinal section
Rigid, dense layer of bone
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Decalcified and stained.