Order Entry
United States
ContactUsLinkComponent
Bluing Liquid
Bluing Liquid
Catalog # 470174-560
CAS Number:  
undefined
Bluing Liquid
Catalog # 470174-560
Supplier Number:  91208
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Bluing Liquid, 8 oz Bottle
  • Cat. No.:
    470174-560
  • Supplier no.:
    91208

Specifications

About this item

Shelf Life (months): Indefinite
Storage: Green

Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.