ContactUsLinkComponent
Biodiesel from New Vegetable Oil Lab Activity
Biodiesel from New Vegetable Oil Lab Activity
Catalog # 470016-730
Supplier:  Avantor
undefined
Biodiesel from New Vegetable Oil Lab Activity
Catalog # 470016-730
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Biodiesel from New Vegetable Oil Lab Activity
  • Environmentally Preferable:
  • No. of lab groups:
    10
  • Cat. No.:
    470016-730
  • Supplier no.:
    WARD470016-730
Specifications
About this item
The grocery store meets the gas station in this innovative, new activity.
  • Ideal for Grades 9-12
  • Activity Demonstrates Green Chemistry Concepts
  • Cost-Effective
  • Efficient
  • Estimated Lab Time: Four Classes, 50 Minutes Each
Students explore green solutions to environmental issues by implementing chemistry techniques to convert new vegetable oil to diesel fuel.
The final products are nontoxic and biodegradable, and the cleaned, dry biodiesel product is road ready for any diesel powered vehicle.
Accessories information: 10 Lab Groups
Ordering information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout. Materials in this lab include vegetable oil, methyl alcohol, potassium hydroxide, a micro burner, a teacher's manual, and student copymasters. Materials that are need but not included beakers, graduated cylinders, thermometers, hotplate/stirrers, electronic balance, and water-proof self-adhesive. This item requires one of the three items: large pipettes & bulbs, separatory funnels, or wash bottles. A fume hood, available separately, is required to perform a portion of this lab activity.