ContactUsLinkComponent
Ward's® Bioassay — Effects of Environmental Toxins Lab Activity
Ward's® Bioassay — Effects of Environmental Toxins Lab Activity
Catalog # 470175-680
Supplier:  Avantor
undefined
Ward's® Bioassay — Effects of Environmental Toxins Lab Activity
Catalog # 470175-680
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Ward's® Bioassay Experiment
  • No. of lab groups:
    5
  • Cat. No.:
    470175-680
  • Supplier no.:
    WARD470175-680
Specifications
About this item
Conduct Reference Toxicity Tests
  • Grow and analyze buttercrunch lettuce
  • Design a bioassay experiment
  • Includes a teacher's guide
Sites of spills and contamination are constantly identified. Help students understand how toxicity tests are performed in areas of environmental distress with this practical lab. Students test the toxicity of salt on the growth of Buttercrunch lettuce seeds, which are easy to culture and require no maintenance between experiments. Applying their results to environmental problems, students then design another bioassay experiment. Includes a teacher’s guide, student copymasters, and materials for five setups.