ContactUsLinkComponent
Bed of Nails Demonstration Model
Bed of Nails Demonstration Model
Catalog # 470004-754
undefined
Bed of Nails Demonstration Model
Catalog # 470004-754
Supplier Number:  47418-00
Specifications
  • Description:
    Bed of Nails Demonstration Model
  • Cat. No.:
    470004-754
  • Supplier no.:
    47418-00
Specifications
About this item
Reveal the Physics of Pressure
Show your students the difference between force and pressure by applying force over a large area. The larger the area, the less likely the balloon is to break. It includes instructions and all materials but the balloons and masses.