Batrachospermum
470194-682
- Description:Batrachospermum (Small)
- No. of students:30
- Cat. No.:470194-682
- Packaging:Tube
- Media:Basic culture solution
- Supplier no.:WARD470194-682
Freshwater. Filamentous. Basic culture solution.
Characterized by reddish plastids (rhodoplasts).