Specifications
- Description:Basic Chromatography Kit
- Cat. No.:470121-696
- Includes:Teacher's Guide
- Supplier no.:15
- No. of uses:30
Specifications
About this item
This kit is a exceptional introduction to chromatography.
- Kit Shows How Chromatography Identifies Makeup of Organic and Inorganic Substances
- Students Compare Effects of Different Solvents
- Required Materials Included
- Chromatography Tubes and Holders Included
- Dyes Included
Students learn how chromatography can identify the makeup of organic and inorganic substances.
They perform experiments to compare the effects of different solvents on the separation of chlorophyll extract and dyes into their constituent components.