Bangia
470179-894
- Description:Bangia (Small)
- No. of students:30
- Cat. No.:470179-894
- Packaging:Tube
- Media:Erdschreiber's medium
- Supplier no.:WARD470179-894
Marine. Filamentous; unbranched thallus. Erdschreiber’s medium.
Characterized by reddish plastids (rhodoplasts).