Orders LinkContactUsLinkComponent
Balancing Chemical Equations: Inquiry-Based Experiment
Supplier:   Avantor
Balancing Chemical Equations: Inquiry-Based Experiment
Supplier:  Avantor
Certificates
Balancing Chemical Equations: Inquiry-Based Experiment
Balancing Chemical Equations: Inquiry-Based Experiment
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Discover Double Replacement Reactions in Microscale.
  • Ideal for reinforcing how to balance a chemical equation
  • Students use knowledge to predict products
  • Aligned to NGSS standards (HS: PS1)
In this engaging experiment kit, developed by high school teacher Alan Hooks, students can observe more than 20 chemical reactions by mixing any two of ten solutions. Students will be very excited to observe the formation of many colorful precipitates, gas bubbles, and solution color changes. After observing these intriguing chemical reactions, students will predict the products, and write the complete balanced chemical equations for the double replacement reactions. Chemical usage is minimized by using spot plates and dropper pipettes. Advanced students can use this experiment kit as an introduction to solubilities, as well as a review of double replacement reactions and predicting products of chemical reactions. Includes solutions of eight salts and two acids, spot plates, pipettes and instructions.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
 : Includes Reaction Plates, Beral Pipettes, Labels, Permanent Markets, Instructions, BaCl2 Solution, CuSO4 Solution, Pb(NO3)2 Solution, KI Solution, AgNO3 Solution, Na2CO3 Solution, and ZnSO4 Solution. Required but not Provided: Erlenmeyer Flasks, Reagent Bottles, Wash Bottles, Waste Containers, and Rinse Water.
Product Family Options
Product Information
  • Includes
    No. of lab groups
    Availability
    Price
  • 470039-268
    Balancing Chemical Equations: Inquiry-Based Experiment
    Balancing Chemical Equations: Inquiry-Based Experiment
    Teacher's Guide and Student Copymaster
    12
    Log in for availability
    Log in for availability
    Each
    Each - $185.59
    $185.59
    Each
       $185.59
  • 470328-441
    Kit with NewPath Digital Resources
    Kit with NewPath Digital Resources
    12
    Discontinued