ContactUsLinkComponent
Ward's® Bacterial Synergism Lab Activity
Ward's® Bacterial Synergism Lab Activity
Catalog # 470030-192
Supplier:  Avantor
undefined
Ward's® Bacterial Synergism Lab Activity
Catalog # 470030-192
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Bacterial Synergism Lab Activity
  • Environmentally Preferable:
  • No. of lab groups:
    6
  • Cat. No.:
    470030-192
Specifications
About this item
Observe a relationship between two organisms that together produce an effect they could not achieve individually.
  • Observe the phenomenon of synergism in your laborat ory
  • Microorganisms included in this set are non-pathogenic
Synergy is the interaction of two or more influences to produce an effect greater than the sum of their individual effects, and students can observe this phenomena between two mutant strains of Serratia marcescens. When cultured separately, each strain is colorless due to a deficiency in the biochemical pathway for pigment production. However, when grown in proximity to each other, each strain completes the other’s biochemical pathway and both are then able to display a red pigment.
Ordering information: This item contains a copupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
Delivery: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Caution: While the organisms in this activity are non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.