Lab-Aids® Bacteria Study Kit
Bacteria Study Kit, 30
Catalog # 470112-506
Supplier: Lab-Aids
: 160
Jump to:
Product Details & Documents
Conduct bacterial experiments in a self-paced activity.
- Prepare and stain bacterial smears
- Observe and identify bacteria
- Time required: one 50 minutes class period
Students conduct the classic Leeuwenhoek bacteria experiment in this individualized, self-paced kit. Paralleling methods performed over 300 years ago, students observe and identify bacteria using a microscope. Students ultimately observe and identify bacteria through a microscope (required, but not included). Bacteria is cultured by peppercorn, hay and bean infusions prepared by the student and then mounted and gram stained on slides.
Included: student worksheets and an instructors manual
Materials needed but not supplied: microscopes
: See datasheet for contents information.
Specifications
- Description:Bacteria Study Kit
- No. of lab groups:30