ContactUsLinkComponent
Ward's® Bacteria Forms Set
Ward's® Bacteria Forms Set
Catalog # 470179-610
Supplier:  Avantor
undefined
Ward's® Bacteria Forms Set
Catalog # 470179-610
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    WARD’S Bacteria Forms Set, Cultures Only
  • Cat. No.:
    470179-610
  • Supplier no.:
    WARD470179-610
Specifications
About this item
Includes Examples of the Three Basic Morphological Types
View all three types of bacteria individually, then compare their different structures, with the selection of specimens in this set. Tube cultures: Bacillus subtilis (rod), Rhodospirillum rubrum (spirillum), and Kocuria rhizophila (coccus). The set of three is available with or without an Explano-Mount, which is a microscope slide with an illustrated guide that highlights all the important information on the slide and the Working with Bacteria manual. Set of three.