Ward's® Live Arcella Culture
Catalog # 470176-436
Supplier: Ward's Science
Specifications
- Description:Arcella vulgaris (Small)
- No. of students:30
- Cat. No.:470176-436
- Packaging:Jar
- Media:Amoeba medium
- Supplier no.:WARD470176-436
Specifications
About this item
Usually found in freshwater and mosses, this organism has a hard umbrella shaped shell or "test", out of which finger-like pseudopodia project.
- Contains a "Test"
- Pseudopodia
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.