ContactUsLinkComponent
Antibody-Antigen Interaction Lab Activity
Antibody-Antigen Interaction Lab Activity
Catalog # 470193-140
Supplier:  Edvotek
undefined
Antibody-Antigen Interaction Lab Activity
Catalog # 470193-140
Supplier:  Edvotek
Supplier Number:  270
Specifications
  • Description:
    Antibody-Antigen Interaction Lab Activity
  • No. of lab groups:
    10
  • Cat. No.:
    470193-140
  • Supplier no.:
    270
Specifications
About this item
Immunodiffusion procedure makes results easy to visualize.
  • Students perform a classic experiment
  • Uses non-human antibodies for safety
The interaction between antigens and antibodies is vital to all aspects of immunology, and this handy activity clearly demonstrates that relationship. Students employ the Ouchterlony Procedure to demonstrate the formation of an antigen-antibody complex through passive diffusion, resulting in an easily observed precipitate in the diffusion medium. For a realistic yet safe experience, the activity utilizes antigens and antibodies from animals; no human components are used. It includes materials for ten setups and instructions.
Ordering information: Includes perishable materials. Please specify delivery date.