ContactUsLinkComponent
Altitude Finder
Altitude Finder
Catalog # 470007-446
Supplier:  Avantor
undefined
Altitude Finder
Catalog # 470007-446
Supplier:  Avantor
Specifications
  • Description:
    One-Piece Clinometer with Trigger
  • Environmentally Preferable:
  • Cat. No.:
    470007-446
  • Supplier no.:
    302232
Specifications
About this item
Durable and Easy to Use
Simply pull the "trigger" to determine the angle to the highest point of an object. Then it's easy to calculate the distance or elevation. The unit can also be placed on an inclined surface to determine the dip angle of rock units. It is constructed of heavy-duty styrene that is lightweight, yet withstands rough use. Size: 11'' L x 6" x 5/8'' W.