BoneClones® Alien Skull
Alien Skull, 2, 22.9 cm, 25.4 cm, 22.9 cm
470029-282
: CC-05
Add A New Twist To Your Comparative Anatomy Lessons
Get your students’ attention and promote lively discussions on the possibility of life on other worlds with this “accurate” representation of an alien skull. Created by a noted special effects artist, this striking skull looks like the real thing! It includes a Plexiglas stand.
- Description:Alien Skull
- Height:22.9 cm
- Length:25.4 cm
- Width:22.9 cm
- No. of parts:2



