Hair Dryer
470015-950
- Description:Hair Dryer
- Input voltage:110 V
- Cat. No.:470015-950
- Supplier no.:3703
An ergonomic handle and two heat/speed settings.
- 1875 watts
- Hang ring
- 5-ft. power cord
For drying glassware, electrostatic equipment, chromatograms and precipitates, and for many other laboratory uses.