ContactUsLinkComponent
Ward's® AGI/NAGT Laboratory Rock Set
Ward's® AGI/NAGT Laboratory Rock Set
Catalog # 470025-288
Supplier:  Avantor
undefined
Ward's® AGI/NAGT Laboratory Rock Set
Catalog # 470025-288
Supplier:  Avantor
Specifications
  • Description:
    Ward's® AGI/NAGT laboratory rock set
  • No. of samples:
    36
  • Cat. No.:
    470025-288
  • Supplier no.:
    WARD470025-288
Specifications
About this item
Specimens To Supplement the Most Widely-Used College Lab Manual for Physical Geology
  • Designed for use with Lab Manual in Phyical Geology
  • Hand made by WARD's Geologists
  • Provides excellent examples of Igneous, Sedimentary and Metamorphic Rocks
Specifically designed to supplement laboratory exercises 4, 5, 6, and 7 of your AGI/NAGT Laboratory Manual in Physical Geology, this unique set of 36 different rocks provides excellent examples of the many key igneous, sedimentary, and metamorphic lithologies.
Identification becomes an interesting hands-on experience when you use Ward's® Rock Set, housed in a compartmented collection box with an accompanying identification list.
Contents: 36 numbered samples, 1 × 1" to 1" × 1¹/₂"
Ordering information: Contents include andesite, arkose, basalt, breccia, chalk, coal (2), conglomerate, coquina, diorite, dolostone, gabbro, gneiss, granite (2), gypsum, lignite, limestone, marble (2), metaconglomerate, obsidian, peridotite (2), phyllite, pumice, quartz, quartzite, rhyolite, sandstone, scoria, serpentinite, shale, schist (2), and slate.