ContactUsLinkComponent
9-Second Flexible Tip Digital Thermometer
9-Second Flexible Tip Digital Thermometer
Catalog # 470160-750
Supplier:  HEALTH SMART SE
undefined
9-Second Flexible Tip Digital Thermometer
Catalog # 470160-750
Supplier:  HEALTH SMART SE
Supplier Number:  15-736-000
Specifications
  • Description:
    9-Second Flexible Tip Digital Thermometer
  • Cat. No.:
    470160-750
  • Supplier no.:
    15-736-000
Specifications
About this item
The soft, flexible tip on this waterproof Fahrenheit thermometer conforms to the shape of the mouth for a safe and comfortable reading.
  • Flexible Mouthpiece for More Comfort
  • Large Display Screen
  • Auto Shut off
  • Waterproof
  • Instructions Provided
The easy-to-read, large display screen emits a tone at peak temperature.
Its auto shut off feature saves power.
Ordering information: This kit includes five probe covers, a storage case, instructions, and a lifetime limited manufacturer's warranty.