ContactUsLinkComponent
40X TAE Electrophoresis Buffer
40X TAE Electrophoresis Buffer
  470203-888
 :  
undefined
40X TAE Electrophoresis Buffer
  470203-888
 :  V4281
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Tris Acetate (TAE), 40X, 1L
  • Cat. No.:
    470203-888
  • Supplier no.:
    V4281
The most commonly used buffer for agarose electrophoresis, this Tris-acetate-EDTA is certified free of Dnase, Rnase, and protease activity. Dilute 1 part in 39 parts deionized water. Store at room temperature. Available to U.S. customers only.