To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:1,10-Phenanthroline Monohydrate
- Size:5 g
- Chemical/physical specifications:Powder
- Cat. No.:470300-000
- Grade:REAG
- Supplier no.:PP0114-5GR
CAS Number: 5144-89-8
Formula Weight: 198.22
Formula: C12H8N2.H2O
Hazard Info: Toxic
Freezing Point (°C): 98-100
Solubility: Water and Alcohol
Synonyms: o-Phenanthroline
Shelf Life (months): 36
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.