ContactUsLinkComponent
DuPont™ Universal Pressure Test Kit for Encapsulated Level A Suits
DuPont™ Universal Pressure Test Kit for Encapsulated Level A Suits
Catalog # 13451-706
Supplier:  DuPont
undefined
DuPont™ Universal Pressure Test Kit for Encapsulated Level A Suits
Catalog # 13451-706
Supplier:  DuPont
Supplier Number:  99081000000001UV
Specifications
  • Description:
    Universal Pressure Test Kit, Input Voltage 85-264VAC @ 47-63 Hz or 120-370VDC
  • Cat. No.:
    13451-706
  • Supplier no.:
    990810000000
Specifications
About this item
This test kit is designed for periodic air pressure testing on all Level A fully encapsulated suits.
  • Compact
  • Lightweight
  • Completely self contained
  • Does not require an external air supply
This compact, lightweight kit is completely self contained, and requires no external air supply.
Certifications: Pressure test kits are used to test Level A garments in accordance with the methods described in ASTM F 1054, "Practice for Pressure Testing of Totally Encapsulated Chemical Protective Suits" and EN464 "Leak-Tightness of Gas-Tight Suits (Internal Pressure Test)".
Caution: Proper decontamination and visual inspection for damage and excessive wear and tear must occur before initiating any pressure testing procedures.