Order Entry
United States
ContactUsLinkComponent
StaticGuard™ Antistatic Reclosable Zip Bags
StaticGuard™ Antistatic Reclosable Zip Bags
Catalog # 82027-726
Supplier:  Minigrip
undefined
StaticGuard™ Antistatic Reclosable Zip Bags
Catalog # 82027-726
Supplier:  Minigrip
Supplier Number:  MGST4P02503

Specifications

  • Thickness:
    4 mil.
  • Environmentally Preferable:
  • Cat. No.:
    82027-726
  • W×L:
    64×76 mm (2.5×3")
  • Supplier no.:
    MGST4P02503

Specifications

About this item

Bags qualify as Type II under MIL-B-81705b entitled barrier material, flexible and electrostatic free.

They will dissipate an induced 5000 V charge to technical zero in accordance with FTS 101C, method 4046.1. Pink.