Order Entry
United States
ContactUsLinkComponent
IGEPAL® CA-720
IGEPAL® CA-720
Catalog # 700014-554
Small Business Enterprise
CAS Number:  
IGEPAL® CA-720
Catalog # 700014-554
Supplier Number:  I1149-500GM
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass, amber
  • Size:
    500 g
  • Cat. No.:
    700014-554
  • Supplier no.:
    I1149-500GM

Specifications

About this item

 :
Density:1.104
Storage Temperature:Ambient
CAS Number:9036-19-5
Formula:C₁₄H₂₁(C₂H₄O)nOH
MDL Number:MFCD00128254