Adapter, Bleed, Ace Glass Incorporated
Catalog # 89064-810
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Bleed Adapter, Bent
- Cat. No.:89064-810
- Supplier no.:9059-12
Specifications
About this item
This adapter comes with a drawn capillary tip for the introduction of gases below the liquid surface.
- Drawn capillary tip
- With hose connection
- Fabricated from borosilicate glass
Adapter can also be used with a #7 Ace-Thred. Outside diameter is 7mm; length is approximately 27cm (10-3/4"). Use with a 3/8-inch I.D. tubing, size D hose connection.