Multineck, 'Mini' #7 Ace-Thred Adapter
Catalog # 89051-514
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Multineck, "Mini' #7 Ace-Thred Adapter
- Cat. No.:89051-514
- Bottom inner [ST] joint size:24/25
- Top outer st joint size:[ST] 24/40
- Supplier no.:5263-17
Specifications
About this item
This adapter has a standard taper inner joint at the bottom, two #7 Ace-Threds and one standard taper outer joint at the top.
- Two Ace-Threds and one [ST] joint at top
- Bottom [ST] joint
- Fabricated from borosilicate glass
Ace-Threds would commonly be used for thermometers or gas inlet tubes, thus leaving joint for condenser, addition funnel, and still head. Standard taper 24/25 medium length joint is compatible with a standard taper 24/40 full length joint.
Ordering information: Supplied complete with two 89051-122 nylon bushings and FETFE 89060-958 o-rings.