Adapter, Thermometer Joint, Offset, Ace Glass Incorporated
89051-320
Product available on GSA Advantage®
Small Business Enterprise
- Description:Offset Thermometer Joint Adapter
- Cat. No.:89051-320
- Supplier no.:5100-10
This adapter features a standard taper thermometer joint at the top and standard taper bottom and side joints.
- Top [ST] thermometer joint
- Bottom [ST] inner joint
- Side [ST] outer joint
- Fabricated from borosilicate glass
Adapter comes with a standard taper 10/30 thermometer joint for a 76mm immersion at the top. Also features a standard taper 24/40 inner bottom joint and standard taper 24/40 outer side joint.