Adapter, Distilling Trap, Ace Glass Incorporated
89051-306
Product available on GSA Advantage®
Small Business Enterprise
- Description:Distilling Trap Adapter
- Environmentally Preferable:
- Cat. No.:89051-306
- Supplier no.:5230-10
The distilling trap adapter is used to prevent carryover of reactions that have a tendency to foam.
- Outlet tube bent at 75° angle
- [ST] 24/40 joint
- Fabricated from borosilicate glass
This distilling trap adapter has an outlet tube bent at a 75 degree angle.