ContactUsLinkComponent
Adapter, Twin, Ace Glass Incorporated
Adapter, Twin, Ace Glass Incorporated
Catalog # 89051-182
Supplier:  Ace Glass
Small Business Enterprise
undefined
Adapter, Twin, Ace Glass Incorporated
Catalog # 89051-182
Supplier:  Ace Glass
Supplier Number:  5034-11
Small Business Enterprise
Specifications
  • Description:
    Twin Adapter
  • Cat. No.:
    89051-182
  • Supplier no.:
    5034-11
Specifications
About this item
This adapter comes with an inner standard taper joint at bottom, one #7 Ace-Thred, and one 10/18 standard taper outer joint, offset at the top.
  • Threaded opening
  • Top Ace-Thred and [ST] outer joints, offset
  • Bottom inner [ST] joint
  • Bushing and o-ring included
  • Fabricated from borosilicate glass
Threaded opening will accept 6.5 to 7mm diameter thermometers, bleed tubes, etc. The 24/25 medium length joint is compatible with a full length 24/40 joint. Supplied complete with 89051-122 nylon bushing and 89060-958 FETFE o-ring.