Specifications
- Temperature range:2...14 °C, ±3 °C (refrigerator),
–20...–30 °C, ±5 °C (freezer) - Capacity:415 - 176 L (14.7 - 6.2 cu.ft.)
- Description:Refrigerator/freezer
- Model:MPR715FPA
- Cat. No.:89230-124
- Electrical:115 V, 60 Hz
- Shelves/drawers:2 full size shelves (55 lbs. per shelf), 3 (33 lbs. per shelf) (refrigerator), 1 full size shelf (22 lbs. per shelf) (freezer)
- Weight:126 kg (277 lbs.)
- Insulation:CFC-Free Rigid Polyurethane, Foamed-In Place
- Exterior W×D×H:900×715×1910 mm (35.4×28.1×75.2")
- Supplier no.:MPR715F
Specifications
About this item
Ideal for biological, reagents, and vaccine storage, the MPR Series biomedical refrigerators with freezers combine high performance refrigeration and control and alarm/monitoring systems with energy-efficient, cost-effective cabinet design.
- Uniformity with forced air circulation
- Sample security with integrated alarm functions
- Safe and secured storage behind a keyed locking door
- Temperature controls and sensors
- Separate refrigeration systems and compressors
- Calibration adjustment through the control panel
Fan-forced air circulation in the refrigerator provides precise temperature uniformity
Separate refrigeration systems and compressors allow differential control of individual refrigerator and freezer compartments, so the freezer section maintains temperatures as low as –30 °C. Comprehensive set point, alarm, monitoring, and diagnostic functions are maintained by the PHCbi-built microprocessor controller with a digital display of all critical functions. A unique cycle defrost (refrigerator only) initiates only as required and maintains internal temperature uniformity without fluctuations. MPR refrigeration systems are designed to assure stored product safety to meet regulatory guidelines for vaccine and biologicals.
Efficient temperature recovery properties allow for minimal temperature fluctuations around set points. Refrigerator evaporator operates above freezing temperatures, which reduces the need for defrost and avoids exposing products to freezing temperatures. Accurate temperature sensors inside the refrigerator and controllers are built to maintain a set temperature within the 2 °C to 8 °C range, and alarms and temp. monitoring quickly announce adverse operating conditions.