Personal Protection (PPE) ANSI Safety First Signs, National Marker
Catalog # 94021-284
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Dimensions:7.6×12.7 cm (3×5")
- Material:PS Vinyl
- Cat. No.:94021-284
- Legend:Remember Wear Safety Equipment
- Supplier no.:SGA7AP
Specifications
About this item
Graphical ANSI Signs are the perfect way to alert your employees and guests when a hazard is present in your facility
Keep your facility safe, buy an ANSI sign today and prevent an accident tomorrow. Each sign header reads, "Safety First".