ContactUsLinkComponent
GC-2014 Gas Chromatograph, Shimadzu
GC-2014 Gas Chromatograph, Shimadzu
Catalog # 89217-152
Supplier:  Avantor
undefined
GC-2014 Gas Chromatograph, Shimadzu
Catalog # 89217-152
Supplier:  Avantor
Specifications
  • Description:
    GC-2014 Gas Chromatograph
  • Cat. No.:
    89217-152
  • Supplier no.:
    220-94687-02
Specifications
About this item
Ideal for routine analysis, the GC-2014 covers a wide range of applications from capillary to packed column analysis
Advanced Flow Controller (AFC) technology comes standard, offering users the precision of AFC, automatic and manual flow or pressure control. AFC provides superior retention time repeatability. Flow rate can be set easily by keypad input while a large LCD screen graphically monitors the flow rate and column inlet pressure. The GC oven is capable of 20 temperature program ramps and hold steps, and features rapid cooling: 300º C to 50º C < 6 minutes. In addition, it provides users with the flexibility to expand their systems for any application by installing up to four redesigned detectors and three injectors.
Ordering information: Installation included.