BioRobotix® PerkinElmer/Packard Tips, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Molecular BioProducts PerkinElmer/Packard pipette tips are designed to fit PerkinElmer/Packard MultiPROBE II HT and HT EX, PlateTrak, MiniTrak and Evolution P-3 workstations.
- Available in nonsterile or sterile formats
Each pipette tip is lot tested during production using a 15-point QC program on the actual workstation for which they are created. They have a low percentage Coefficient of Variance (%CV). The pipette tips, racks, and lids reduce static charges so tips pick up straight and eject perfectly each time.
They are also available with ART® filter technology and low retention design for critical samples, providing increased precision for the toughest multi-dispensing assays.
Specifications
- Catalog No:
- 72830-514
- 72830-488
- 72830-486
- 72830-484
- 72830-506
- 72830-345
- 89108-098
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)