Coil Reflux Condenser, Microscale, Ace Glass Incorporated
Catalog # 89075-170
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Condenser, Coil Reflux, Microscale
- Cat. No.:89075-170
- Supplier no.:9569-24
Specifications
About this item
High efficiency coil condenser especially suited for vacuum applications
Coil length is approximately 70 mm. The top joint is a combination cap, thread and standard taper outer joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external 20-400 thread to facilitate an o-ring, and an open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. Bottom is standard taper 14/10 inner joint. Fabricated from borosilicate glass.
Ordering information: Comes complete with open-top phenolic cap and o-ring.