Condenser, Allihn
89083-322
Product available on GSA Advantage®
Small Business Enterprise
- Description:Condenser, Allihn
- Cat. No.:89083-322
- Top [ST] joint size:[ST] 35/25
- Jacket length:400 mm
- Supplier no.:5945-65
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with ball joint at bottom and socket joint [SJ] at top
Fabricated from borosilicate glass. Size D glass hose connections, for use with 3/8-inch I.D. tubing.