ContactUsLinkComponent
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
Catalog # 82021-306
Supplier:  G-Biosciences
undefined
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
Catalog # 82021-306
Supplier:  G-Biosciences
Supplier Number:  786-140
Specifications
  • Description:
    GELPLATE-clean™ Spray
  • Size:
    2 x 250 mL
  • Cat. No.:
    82021-306
  • Supplier no.:
    786-140
Specifications
About this item
G-Biosciences' GELPLATE-clean™ has been specifically formulated for cleaning electrophoresis gel plates. Clean your electrophoresis plates with GELPLATE-clean™ to prevent poor electrophoretic band migration, band distortions, and poor image development.
G-Biosciences' GELPLATE-clean™ removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. The solution is suitable for both sequencing and protein electrophoresis gel plates. With GELPLATE-clean™, simply spray your electrophoresis plates and then wipe clean.
Ordering information: Refill solutions are offered separately. Accessories available.
Packaging: Supplied in two easy-to-use 250 mL spray bottles.