Anti-LRRK1 Rabbit Polyclonal Antibody
Catalog # 60943-024
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine-rich Repeat serine/threonine-protein kinase 1
- Antigen Symbol:LRRK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Epitope:N-Terminal (100–200 aa)
- Antigen Synonyms:Roco1|RIPK6
- Cat. No.:60943-024
- Supplier no.:AP7098A
Specifications
About this item
Primary Rabbit Anti-LRRK1 (N-term) Reacts with Human
Type: Primary
Antigen: LRRK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (100–200 aa)
Host: Rabbit
Isotype:
Reactivity: Human