To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Tumor Necrosis Factor Alpha
- Antigen Symbol:TNFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Cross Adsorption:No
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:21926
- Antigen Synonyms:tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This antibody was produced from whole rabbit serum prepared by repeated immunizations with recombinant mouse TNFa produced in E.coli.
- Tested Applications:Radio Immunoassay
- Cat. No.:RL210-401-321
- Supplier no.:210-401-321
This antibody against Mouse TNFa has been tested for use in neutralizations, ELISA, immunochemistry, radioimmunoassay, immunoprecipitation, immunochemistry and immunoblotting. It recognizes the 17,000 MW TNFa.
Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse