Order Entry
United States
ContactUsLinkComponent
Anti-ampC Rabbit Polyclonal Antibody
Anti-ampC Rabbit Polyclonal Antibody
Catalog # RL100-4175
Anti-ampC Rabbit Polyclonal Antibody
Catalog # RL100-4175
Supplier Number:  100-4175
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Penicillinase
  • Antigen Symbol:
    Penicillinase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    E. cloacae
  • Western Blot:
    Yes
  • Size:
    2 mL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Gene ID:
    9125732
  • Storage Buffer:
    0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage Temperature:
    –20 °C
  • Concentration:
    90 mg/ml by Refractometry
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Penicillinase (Enterobacter cloacae)
  • Purification:
    delipidation and defibrination
  • Cat. No.:
    RL100-4175
  • Supplier no.:
    100-4175

Specifications

About this item

Anti-PENICILLINASE Antibody has been assayed against 1.0 µg of Penicillinase (Enterobacter cloacae) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ampC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Enterobacter cloacae