Order Entry
United States
ContactUsLinkComponent
Anti-Luciferase Rabbit Monoclonal Antibody [clone: 17D10]
Anti-Luciferase Rabbit Monoclonal Antibody [clone: 17D10]
Catalog # RL100-401-137
undefined
Anti-Luciferase Rabbit Monoclonal Antibody [clone: 17D10]
Catalog # RL100-401-137
Supplier Number:  100-401-137

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Luciferase
  • Antigen Symbol:
    Luciferase
  • Clonality:
    Monoclonal
  • Clone:
    9B10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Photobacterium fischerii
  • Size:
    2 mL
  • Cross Adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Gene ID:
    3280269
  • Storage Buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage Temperature:
    –20 °C
  • Concentration:
    90 mg/ml by Refractometry
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Luciferase (Photobacterium fischerii)
  • Purification:
    delipidation and defibrination
  • Cat. No.:
    RL100-401-137
  • Supplier no.:
    100-401-137

Specifications

About this item

Primary Rabbit Anti-LUCIFERASE (Photobacterium fischerii) Reacts with Pan

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Luciferase
Clonality: Monoclonal
Clone: 17D10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan